missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNAH2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564309
This item is not returnable.
View return policy
Description
Immunogen sequence: PGEWENACNE MQRMLIVRSL RQDRVAFCVT SFIITNLGSR FIEPPVLNMK SVLEDSTPRS PLVFILSPGV DPTSALL Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 95%.
Force generating protein of respiratory cilia. Produces force towards the minus ends of microtubules. Dynein has ATPase activity; the force-producing power stroke is thought to occur on release of ADP. Involved in sperm motility; implicated in sperm flagellar assembly.
Specifications
| DNAH2 | |
| Polyclonal | |
| Unconjugated | |
| DNAH2 | |
| axonemal beta dynein heavy chain 2; Ciliary dynein heavy chain 2; DNAH2; DNAHC2; DNHD3; dynein axonemal heavy chain 2; dynein heavy chain 2, axonemal; Dynein heavy chain domain-containing protein 3; dynein, axonemal, heavy chain 2; dynein, axonemal, heavy polypeptide 2; KIAA1503 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 146754 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9P225 | |
| DNAH2 | |
| Recombinant protein corresponding to Human DNAH2. Recombinant protein control fragment (Product #RP-103886). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction