missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAH17 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £513.00
Specifications
| Antigen | DNAH17 |
|---|---|
| Dilution | Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18645206
|
Novus Biologicals
NBP2-62702-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18696036
|
Novus Biologicals
NBP2-62702 |
100 μg |
£513.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DNAH17 Polyclonal antibody specifically detects DNAH17 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| DNAH17 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| axonemal dynein heavy chain, axonemal dynein heavy chain-like protein 1, ciliary dynein heavy chain 17, ciliary dynein heavy chain-like protein 1, DNAHL1, DNEL2, dynein heavy chain 17, axonemal, dynein light chain 2, axonemal, dynein, axonemal, heavy chain 17, dynein, axonemal, heavy chain like 1, dynein, axonemal, heavy like 1, dynein, axonemal, heavy polypeptide 17, FLJ40457, MGC132767, MGC138489 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: PWKDYVIYIDDMVLDEFDQFIRKSLSFLMDNMVIDESIAPLFEIRMELDEDGLTFNPTLEVGSDRGFLALIEGLVNDIYNVARLIPRLAKDRMNYKMD | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol | |
| 8632 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title