missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNAH12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570820
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Dog: 79%; Horse: 79%; Human: 100%; Mouse: 93%; Rat: 93%.
DNAH12 gene ontology annotations related to this gene include microtubule-based movement.
Specifications
| DNAH12 | |
| Polyclonal | |
| Unconjugated | |
| DNAH12 | |
| 4921531P07Rik; axonemal beta dynein heavy chain 12; Axonemal dynein heavy chain 12-like protein; Axonemal dynein heavy chain 7-like protein; axonemal dynein heavy chain isotype3; Bm259; ciliary dynein heavy chain 12; DHC3; DLP12; Dnah12; DNAH12L; DNAH7L; DNAHC12; Dnahc3; Dnahc7c; Dnahc7l; DNHD2; dynein axonemal heavy chain 12; dynein heavy chain 12, axonemal; dynein heavy chain 12, axonemal-like; Dynein heavy chain 7-like, axonemal; dynein heavy chain domain 2; dynein heavy chain domain-containing protein 2; dynein, axonemal, heavy chain 12; dynein, axonemal, heavy polypeptide 12; dynein, heavy chain-5; Gm284; Gm74; Gm907; Hdhc3; HL19; HL-19 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 201625 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q6ZR08 | |
| DNAH12 | |
| synthetic peptide directed towards the N-terminal region of human DNAH12. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction