missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNAH11 Monoclonal Antibody (C5), Invitrogen™

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA543853
This item is not returnable.
View return policy
Description
This gene encodes a ciliary outer dynein arm protein and is a member of the dynein heavy chain family. It is a microtubule-dependent motor ATPase and has been reported to be involved in the movement of respiratory cilia. Mutations in this gene have been implicated in causing Kartagener Syndrome (a combination of situs inversus totalis and Primary Ciliary Dyskinesia (PCD), also called Immotile Cilia Syndrome 1 (ICS1)) and male sterility. [provided by RefSeq, Mar 2013].
Specifications
| DNAH11 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q96DT5 | |
| DNAH11 | |
| Leu3896-Asn4122 (Accession Q96DT5), with N-terminal His Tag. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| C5 | |
| Unconjugated | |
| DNAH11 | |
| axonemal beta dynein heavy chain 11; axonemal dynein heavy chain 11; CILD7; ciliary dynein heavy chain 11; DNAH11; DNAHBL; DNAHC11; DNHBL; DPL11; dynein axonemal heavy chain 11; dynein heavy chain 11, axonemal; dynein, axonemal, heavy chain 11; dynein, axonemal, heavy polypeptide 11; dynein, ciliary, heavy chain 11 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 8701 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction