missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAH11 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-49883
This item is not returnable.
View return policy
Description
DNAH11 Polyclonal specifically detects DNAH11 in Human, Mouse, Bovine, Canine, Rhesus Macaque samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
| DNAH11 | |
| Polyclonal | |
| Immunohistochemistry 1:200, Immunohistochemistry-Paraffin 1:200, Peptide ELISA detection limit 1:32000 | |
| NP_003768.2 | |
| DNAH11 | |
| Peptide with sequence NVDEICEFNLNQ corresponding to internal region according to NP_003768.2. | |
| 0.1 mg | |
| Primary | |
| This antibody recognizes and epitope within the internal region.This antibody is specific to the internal region. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| axonemal, Axonemal beta dynein heavy chain 11, CILD7Dnahc11, Ciliary dynein heavy chain 11, DNAHBLFLJ30095, DNAHC11FLJ37699, DNHBL, dynein, axonemal, heavy chain 11, dynein, axonemal, heavy polypeptide 11, dynein, ciliary, heavy chain 11 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8701 | |
| Human, Mouse, Bovine, Canine, Monkey, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction