missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNAAF3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571524
This item is not returnable.
View return policy
Description
DNAAF3 Polyclonal Antibody for Western Blot, IHC (P)
The protein encoded by this gene is required for the assembly of axonemal inner and outer dynein arms and plays a role in assembling dynein complexes for transport into cilia. Defects in this gene are a cause of primary ciliary dyskinesia type 2 (CILD2). Several transcript variants encoding different isoforms have been found for this gene.
Specifications
| DNAAF3 | |
| Polyclonal | |
| Unconjugated | |
| DNAAF3 | |
| 6030429G01Rik; b2b1739Clo; C19orf51; CILD2; DAB1; DNAAF3; dynein; dynein (axonemal) assembly factor 3; dynein assembly factor 3, axonemal; dynein axonemal assembly factor 3; dynein, axonemal assembly factor 3; dynein, axonemal, assembly factor 3; PCD; PF22; UPF0470 protein C19orf51; UPF0470 protein C19orf51 homolog | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 352909, 436022 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q3UYV8, Q8N9W5 | |
| DNAAF3 | |
| KLH conjugated synthetic peptide between 181-210 amino acids from the Central region of human C19orf51. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction