missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNAAF3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566004
This item is not returnable.
View return policy
Description
Immunogen sequence: FVLENNLEAVA RHMLIFSLAL EEPEKMGLQE RSETFLEVWG NALLRPPVAA FVRAQADLLA HLVPEPDRLE EQLPWLSLRA LKFRERDALE AVFRFWA Highest antigen sequence identity to the following orthologs - mouse 90%, rat 89%.
The protein encoded by this gene is required for the assembly of axonemal inner and outer dynein arms and plays a role in assembling dynein complexes for transport into cilia. Defects in this gene are a cause of primary ciliary dyskinesia type 2 (CILD2). Several transcript variants encoding different isoforms have been found for this gene.
Specifications
| DNAAF3 | |
| Polyclonal | |
| Unconjugated | |
| DNAAF3 | |
| 6030429G01Rik; b2b1739Clo; C19orf51; CILD2; DAB1; DNAAF3; dynein; dynein (axonemal) assembly factor 3; dynein assembly factor 3, axonemal; dynein axonemal assembly factor 3; dynein, axonemal assembly factor 3; dynein, axonemal, assembly factor 3; PCD; PF22; UPF0470 protein C19orf51; UPF0470 protein C19orf51 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 352909 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N9W5 | |
| DNAAF3 | |
| Recombinant Human DNAAF3. Recombinant protein control fragment (Product #RP-106691). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction