missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNAAF2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA544697
This item is not returnable.
View return policy
Description
Peptide sequence: MFSQYAEELT DPENRRRYEA EITALERERG VEVRFVHPEP GHVLRTSLDG Sequence homology: Cow: 100%; Dog: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%.
The function of this protein remains unknown.
Specifications
| DNAAF2 | |
| Polyclonal | |
| Unconjugated | |
| DNAAF2 | |
| 1110034A24Rik; 2810020C19Rik; AV032410; C10H14orf104; C14orf104; CILD10; DNAAF2; dynein (axonemal) assembly factor 2; dynein assembly factor 2, axonemal; dynein axonemal assembly factor 2; dynein, axonemal assembly factor 2; dynein, axonemal, assembly factor 2; kintoun; kintoun homolog; Ktu; mknt; PF13; protein kintoun; RGD1310311; TU54 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 55172 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NVR5 | |
| DNAAF2 | |
| Synthetic peptide directed towards the N-terminal of human C14orf104 (aa 35-84). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction