missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNAAF2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551766
This item is not returnable.
View return policy
Description
Immunogen sequence: LQGKEERVNE ESHLTEKEYI EHCNTPTTDS DSSIAVKALQ IDSFGLVTCF QQESLDVSQM ILGKSQQPES KMQSEFIKEK SATCSNEEKG NLNESVITEE KETDGDHLSS LLNKTTVHNI PGFDSIKETN MQDGSVQVIK Highest antigen sequence identity to the following orthologs: Mouse - 51%, Rat - 57%.
The function of this protein remains unknown.
Specifications
| DNAAF2 | |
| Polyclonal | |
| Unconjugated | |
| DNAAF2 | |
| 1110034A24Rik; 2810020C19Rik; AV032410; C10H14orf104; C14orf104; CILD10; DNAAF2; dynein (axonemal) assembly factor 2; dynein assembly factor 2, axonemal; dynein axonemal assembly factor 2; dynein, axonemal assembly factor 2; dynein, axonemal, assembly factor 2; kintoun; kintoun homolog; Ktu; mknt; PF13; protein kintoun; RGD1310311; TU54 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55172 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NVR5 | |
| DNAAF2 | |
| Recombinant protein corresponding to Human DNAAF2. Recombinant protein control fragment (Product #RP-89061). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction