missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNA2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568167
This item is not returnable.
View return policy
Description
Predicted to react with Mouse samples.
DNA2 is a conserved helicase/nuclease involved in themaintenance of mitochondrial and nuclear DNA stability.
Specifications
| DNA2 | |
| Polyclonal | |
| Unconjugated | |
| DNA2 | |
| DNA replication ATP-dependent helicase DNA2; DNA replication ATP-dependent helicase/nuclease DNA2; DNA replication ATP-dependent helicase-like homolog; DNA replication helicase 2 homolog; DNA replication helicase 2 homolog (yeast); DNA replication helicase 2-like; DNA replication helicase/nuclease 2; DNA replication nuclease DNA2; DNA2; DNA2L; DNA2-like helicase; E130315B21Rik; hDNA2; Kiaa0083; LOW QUALITY PROTEIN: DNA replication ATP-dependent helicase/nuclease DNA2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1763, 327762 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P51530, Q6ZQJ5 | |
| DNA2 | |
| Synthesized peptide derived from human DNA2. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction