Learn More
DNA2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21599-1-AP-20UL
 View more versions of this product
Product Code. 16891195
  • £135.00 / 20µL
Description
This antibody is specific to DNA2.
DNA2 is a conserved helicase/nuclease involved in themaintece of mitochondrial and nuclear DNA stability.
Specifications
Specifications
DNA2
Polyclonal
Unconjugated
DNA2
DNA2, DNA2L, KIAA0083
Rabbit
Antigen Affinity Chromatography
RUO
1763
-20°C
Liquid
Immunoprecipitation, Immunohistochemistry (Paraffin)
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P51530
DNA2
DNA2 Fusion Protein Ag16206
20 μL