Learn More
Fisherbrand™ DNA Sequencing Systems: Combs for Discontinued Model FB-SEQ-2045
DNA Sequencing Systems Combs for discontinued model FB-SEQ-2045
Brand: Fisherbrand™
Code : 04
Additional Details : Weight : 0.22000kg
Description
For use with discontinued model FB-SEQ-2045
Specifications
Owl s4s aluminum backed sequencer system | |
73 | |
3.7μL |
Sharktooth Micro-Well Comb | |
0.4 mm |