Learn More
Labconco DNA Rotor - CentriVap Pro
Product Code. 30533441 Shop All Labconco Corporation Products
Change view
Click to view available options
Unit Size:
Each
Material (Coating):
PTFE
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size Material (Coating)
30533441 Each -
30533434 Each PTFE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30533441 Supplier Labconco Supplier No. 7555700
  • ONLINE EXCLUSIVE PRICE
    £333.00
    £326.41 / Pack of 1
    Save £6.59
    1% Off
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Rotors for use with Labconco™ CentriVap™ Pro Vacuum Concentrators. Stainless steel and PTFE coated options available.
  • Compatible with CentriVap Pro Vacuum Concentrators
  • Stackable design
  • Stainless steel material\
  • Optional PTFE coated
Specifications
Material Stainless Steel
Type Rotor
Rotor System CentriVap Pro
Dimensions (L x W x H) 26.42 x 22.86 x 6.6 cm (10.4 x 9 x 2.6 in.)