missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA-RNA Hybrid Antibody (S9.6), Novus Biologicals™
Mouse Monoclonal Antibody
£195.00 - £368.00
Specifications
| Antigen | DNA-RNA Hybrid |
|---|---|
| Clone | S9.6 |
| Concentration | 0.2 mg/ml |
| Applications | ChIP Assay, Flow Cytometry, Immunoprecipitation, Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30300934
|
Novus Biologicals
NBP3-44628-100ug |
100 μg |
£368.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30303464
|
Novus Biologicals
NBP3-44628-20ug |
20 μg |
£195.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DNA-RNA Hybrid Monoclonal antibody specifically detects DNA-RNA Hybrid in All samples. It is validated for ChIP assay,Flow Cytometry,Immunoprecipitation,Immunocytochemistry/ ImmunofluorescenceSpecifications
| DNA-RNA Hybrid | |
| 0.2 mg/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| IgG2a κ | |
| Protein A purified |
| S9.6 | |
| ChIP Assay, Flow Cytometry, Immunoprecipitation, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| All species | |
| DNA-RNA heteropolymer duplex prepared by transcription of phi X174 single-stranded DNA with DNA-dependent RNA polymerase | |
| Primary | |
| Store at 4⪚C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title