missing translation for 'onlineSavingsMsg'
Learn More

DNA Primase small subunit Antibody [DyLight 755], Novus Biologicals™

Product Code. 30763972 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30763972 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30763972 Supplier Novus Biologicals Supplier No. NBP346896IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DNA Primase small subunit Polyclonal antibody specifically detects DNA Primase small subunit in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNA Primase small subunit
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias DNA primase 1, DNA primase 49 kDa subunit, DNA primase small subunit, DNA primase subunit 48, EC 2.7.7, EC 2.7.7.-, MGC12308, p49, primase p49 subunit, primase polypeptide 1, 49kDa, primase, DNA, polypeptide 1 (49kDa)
Host Species Rabbit
Immunogen primase, DNA, polypeptide 1(49kDa). (Uniprot# P49642)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Lipid and Metabolism, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5557
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.