missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA Primase large subunit Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-84798-0.1ML
This item is not returnable.
View return policy
Description
DNA Primase large subunit Polyclonal specifically detects DNA Primase large subunit in Human samples. It is validated for Western Blot.
Specifications
| DNA Primase large subunit | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| dJ422B11.1.1, DNA primase 58 kDa subunit, DNA primase subunit p58, EC 2.7.7, EC 2.7.7.-, MGC75142, p58, polypeptide 2A, p58, PRIM2ADNA primase large subunit, primase polypeptide 2A, 58kDa, primase, DNA, polypeptide 2 (58kDa), primase, polypeptide 2A (58kD), primase, polypeptide 2A, 58kDa | |
| The immunogen is a synthetic peptide directed towards the middle region of human DNA Primase large subunit | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 5558 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction