missing translation for 'onlineSavingsMsg'
Learn More

DNA Primase large subunit Antibody [PE], Novus Biologicals™

Product Code. 30764590 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30764590 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30764590 Supplier Novus Biologicals Supplier No. NBP346895PE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DNA Primase large subunit Polyclonal antibody specifically detects DNA Primase large subunit in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNA Primase large subunit
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate PE
Formulation PBS
Gene Alias dJ422B11.1.1, DNA primase 58 kDa subunit, DNA primase subunit p58, EC 2.7.7, EC 2.7.7.-, MGC75142, p58, polypeptide 2A, p58, PRIM2ADNA primase large subunit, primase polypeptide 2A, 58kDa, primase, DNA, polypeptide 2 (58kDa), primase, polypeptide 2A (58kD), primase, polypeptide 2A, 58kDa
Host Species Rabbit
Immunogen primase, DNA, polypeptide 2(58kDa). (Uniprot# P49643)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5558
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.