missing translation for 'onlineSavingsMsg'
Learn More

DNA polymerase sigma Antibody (2F8), Novus Biologicals™

Product Code. 18357389 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18357389 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18357389 Supplier Novus Biologicals Supplier No. H00011044M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DNA polymerase sigma Monoclonal antibody specifically detects DNA polymerase sigma in Human samples. It is validated for Western Blot, ELISA, KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNA polymerase sigma
Applications Western Blot, ELISA, KnockDown
Classification Monoclonal
Clone 2F8
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_008930
Gene Alias DNA polymerase kappa, DNA polymerase sigma, EC 2.7.7.7, LAK-1TUTase 5, PAP associated domain containing 7, POLKTRF41, POLSTUTASE5, polymerase (DNA directed) sigma, polymerase (DNA-directed) sigma, Terminal uridylyltransferase 5, Topoisomerase-related function protein 4-1, TRF4-1LAK1, TRF4PAP-associated domain-containing protein 7
Host Species Mouse
Immunogen POLS (NP_008930, 2 a.a. ~ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. SPCPEEAAMRREVVKRIETVVKDLWPTADVQIFGSFSTGLYLPTSDIDLVVFGKWERPPLQLLEQALRKHNVAEPCSIKVLDKATVPIIKLTDQETEVKVDISFNMETG
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11044
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.