missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA polymerase mu Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-16189
This item is not returnable.
View return policy
Description
DNA polymerase mu Polyclonal specifically detects DNA polymerase mu in Human samples. It is validated for Western Blot.
Specifications
| DNA polymerase mu | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| DNA polymerase mu, DNA-directed DNA/RNA polymerase mu, EC 2.7.7.7, FLJ35482, Pol iota, Pol Mu, polmu, polymerase (DNA directed), mu, polymerase (DNA-directed), mu, Tdt-N, Terminal transferase | |
| Rabbit | |
| 55 kDa | |
| 0.1 mL | |
| DNA Repair, DNA replication Transcription Translation and Splicing | |
| 27434 | |
| Human | |
| IgG |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q9NP87 | |
| POLM | |
| Recombinant protein encompassing a sequence within the center region of human DNA polymerase mu. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Expected to cross react based on sequence identity: Bovine (82%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction