missing translation for 'onlineSavingsMsg'
Learn More

DNA Polymerase Kappa Antibody, DyLight 650, Novus Biologicals™

Product Code. 18426964 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18426964 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18426964 Supplier Novus Biologicals Supplier No. NBP106559C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DNA Polymerase Kappa Polyclonal antibody specifically detects DNA Polymerase Kappa in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNA Polymerase Kappa
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias DINB protein, DINPPOLQDINB1EC 2.7.7.7, DNA polymerase kappa, polymerase (DNA directed) kappa
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 825 and 870 of human DNA polymerase kappa using the numbering given in entry NP_057302.1
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51426
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.