missing translation for 'onlineSavingsMsg'
Learn More

DNA Polymerase iota Antibody (OTI5A4) [Alexa Fluor™ 700], Novus Biologicals™

Product Code. 30688612 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30688612 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30688612 Supplier Novus Biologicals Supplier No. NBP344665AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DNA Polymerase iota Monoclonal antibody specifically detects DNA Polymerase iota in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNA Polymerase iota
Applications Western Blot
Classification Monoclonal
Clone OTI5A4
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias DNA polymerase iota, EC 2.7.7.7, Eta2, polymerase (DNA directed) iota, RAD30 homolog B, RAD30Bpolymerase (DNA-directed), iota, RAD3OB
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 458-740 of human POLI (NP_009126) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Research, DNA Polymerases, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 11201
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.