missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA Polymerase iota Antibody (OTI2C10), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
£195.00 - £417.00
Specifications
| Antigen | DNA Polymerase iota |
|---|---|
| Clone | OTI2C10 |
| Dilution | Western Blot 1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30306256
|
Novus Biologicals
NBP3-44872-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30306699
|
Novus Biologicals
NBP3-44872-30ul |
30 μL |
£195.00
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DNA Polymerase iota Monoclonal antibody specifically detects DNA Polymerase iota in Human samples. It is validated for Western BlotSpecifications
| DNA Polymerase iota | |
| Western Blot 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| DNA polymerase iota, EC 2.7.7.7, Eta2, polymerase (DNA directed) iota, RAD30 homolog B, RAD30Bpolymerase (DNA-directed), iota, RAD3OB | |
| Human recombinant protein fragment corresponding to amino acids 458-740 of human POLI (NP_009126) produced in E.coli. | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
| OTI2C10 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Chromatin Research, DNA Polymerases, DNA Repair | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| 11201 | |
| IgG1 | |
| >95% | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title