missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA Polymerase iota Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92404-0.02ml
This item is not returnable.
View return policy
Description
DNA Polymerase iota Polyclonal antibody specifically detects DNA Polymerase iota in Human, Mouse samples. It is validated for Western Blot
Specifications
| DNA Polymerase iota | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DNA polymerase iota, EC 2.7.7.7, Eta2, polymerase (DNA directed) iota, RAD30 homolog B, RAD30Bpolymerase (DNA-directed), iota, RAD3OB | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 50-350 of human POLI (NP_009126.2). ASSRVIVHVDLDCFYAQVEMISNPELKDKPLGVQQKYLVVTCNYEARKLGVKKLMNVRDAKEKCPQLVLVNGEDLTRYREMSYKVTELLEEFSPVVERLGFDENFVDLTEMVEKRLQQLQSDELSAVTVSGHVYNNQSINLLDVLHIRLLVGSQIAAEMREAMYNQLGLTGCAGVASNKLLAKLVSGVFKPNQQTVLLPESCQHLIHSLNHIKEIPGIGYKTAKCLEALGINSVRDLQTFSPKILEKELGISVAQRIQKLSFGEDNSPVILSGPPQSFSEEDSFKKCSSEVEAKNKIEELL | |
| 0.02 mL | |
| Chromatin Research, DNA Polymerases, DNA Repair | |
| 11201 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction