missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA Polymerase gamma Rabbit anti-Human, Mouse, Rat, Clone: 9V9D9, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | DNA Polymerase gamma |
|---|---|
| Clone | 9V9D9 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18352797
|
Novus Biologicals
NBP3-15460-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18391414
|
Novus Biologicals
NBP3-15460-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DNA Polymerase gamma Monoclonal antibody specifically detects DNA Polymerase gamma in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| DNA Polymerase gamma | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| DNA Polymerases, DNA Repair | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 5428 | |
| IgG | |
| Affinity purified |
| 9V9D9 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DNA polymerase subunit gamma-1, EC 2.7.7.7, FLJ27114, MDP1, MIRAS, Mitochondrial DNA polymerase catalytic subunit, MTDPS4A, PolG, catalytic subunit, POLG1MTDPS4B, PolG-alpha, POLGAPEO, polymerase (DNA directed), gamma, SANDO, SCAE | |
| A synthetic peptide corresponding to a sequence within amino acids 1140-1239 of human DNA Polymerase gamma (P54098). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title