missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA Polymerase gamma Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-52300
This item is not returnable.
View return policy
Description
DNA Polymerase gamma Polyclonal antibody specifically detects DNA Polymerase gamma in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry
Specifications
| DNA Polymerase gamma | |
| Polyclonal | |
| Unconjugated | |
| 1X PBS (pH 7.0), 20% Glycerol | |
| DNA polymerase subunit gamma-1, EC 2.7.7.7, FLJ27114, MDP1, MIRAS, Mitochondrial DNA polymerase catalytic subunit, MTDPS4A, PolG, catalytic subunit, POLG1MTDPS4B, PolG-alpha, POLGAPEO, polymerase (DNA directed), gamma, SANDO, SCAE | |
| Recombinant protein fragment containing a sequence corresponding to a region within amino acids 829 and 1201 of DNA Polymerase gamma. | |
| 0.05 mL | |
| DNA Polymerases, DNA Repair | |
| 5428 | |
| Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.67 mg/mL | |
| Western Blot 1:1000-1:10000, Immunohistochemistry 10 μg/mL, Immunocytochemistry/Immunofluorescence 1:100 - 1:200, Immunohistochemistry-Paraffin 10 μg/mL, Immunocytochemistry 1:100-1:200 | |
| P54098 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction