missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA Polymerase epsilon subunit 3 Mouse anti-Human, Clone: PCRP-POLE3-2F10, Novus Biologicals™
Mouse Monoclonal Antibody
£377.00
Specifications
| Antigen | DNA Polymerase epsilon subunit 3 |
|---|---|
| Clone | PCRP-POLE3-2F10 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 1 to 2 μg/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunohistochemistry-Paraffin, Protein Array |
| Applications | Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Peptide Array |
Description
DNA Polymerase epsilon subunit 3 Monoclonal specifically detects DNA Polymerase epsilon subunit 3 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Protein Array.Specifications
| DNA Polymerase epsilon subunit 3 | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Chromatin Research, DNA Polymerases, DNA Repair | |
| 10 mM PBS with 0.05% BSA | |
| 54107 | |
| IgG1 | |
| Protein A or G purified |
| PCRP-POLE3-2F10 | |
| Flow Cytometry 1 to 2 μg/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunohistochemistry-Paraffin, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Arsenic-transactivated protein, asTP, CHARAC17arsenic transactivated protein, CHRAC-17, CHRAC17Ybl1, Chromatin accessibility complex 17 kDa protein, DNA polymerase epsilon p17 subunit, DNA polymerase epsilon subunit 3, DNA polymerase epsilon subunit p17, DNA polymerase II subunit 3, EC 2.7.7.7, histone fold protein CHRAC17, HuCHRAC17, p17, polymerase (DNA directed), epsilon 3 (p17 subunit), YBL1 | |
| Recombinant full-length human DNA Polymerase epsilon subunit 3 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title