missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA Polymerase epsilon subunit 3 Antibody (PCRP-POLE3-3D3) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14143JF669
This item is not returnable.
View return policy
Description
DNA Polymerase epsilon subunit 3 Monoclonal antibody specifically detects DNA Polymerase epsilon subunit 3 in Human samples. It is validated for Flow Cytometry,ELISA,Immunoprecipitation,Protein Array,Immunocytochemistry/ Immunofluorescence
Specifications
| DNA Polymerase epsilon subunit 3 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| Arsenic-transactivated protein, asTP, CHARAC17arsenic transactivated protein, CHRAC-17, CHRAC17Ybl1, Chromatin accessibility complex 17 kDa protein, DNA polymerase epsilon p17 subunit, DNA polymerase epsilon subunit 3, DNA polymerase epsilon subunit p17, DNA polymerase II subunit 3, EC 2.7.7.7, histone fold protein CHRAC17, HuCHRAC17, p17, polymerase (DNA directed), epsilon 3 (p17 subunit), YBL1 | |
| Recombinant full-length human DNA Polymerase epsilon subunit 3 protein (Uniprot: Q9NRF9) | |
| 0.1 mL | |
| Chromatin Research, DNA Polymerases, DNA Repair | |
| 54107 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry, ELISA, Immunoprecipitation, Protein Array, Immunocytochemistry/Immunofluorescence | |
| PCRP-POLE3-3D3 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction