missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA Polymerase epsilon subunit 3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£186.00 - £360.00
Specifications
| Antigen | DNA Polymerase epsilon subunit 3 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, Knockout Validated |
| Applications | Western Blot, Gene Knock-Out |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18675510
|
Novus Biologicals
NBP2-92951-0.02ml |
0.02 mL |
£186.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18677810
|
Novus Biologicals
NBP2-92951-0.1ml |
0.1 mL |
£360.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DNA Polymerase epsilon subunit 3 Polyclonal antibody specifically detects DNA Polymerase epsilon subunit 3 in Human, Mouse samples. It is validated for Western Blot, Gene Knock-OutSpecifications
| DNA Polymerase epsilon subunit 3 | |
| Western Blot, Gene Knock-Out | |
| Unconjugated | |
| Rabbit | |
| Chromatin Research, DNA Polymerases, DNA Repair | |
| PBS with 50% glycerol, pH7.3. | |
| 54107 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, Knockout Validated | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| Arsenic-transactivated protein, asTP, CHARAC17arsenic transactivated protein, CHRAC-17, CHRAC17Ybl1, Chromatin accessibility complex 17 kDa protein, DNA polymerase epsilon p17 subunit, DNA polymerase epsilon subunit 3, DNA polymerase epsilon subunit p17, DNA polymerase II subunit 3, EC 2.7.7.7, histone fold protein CHRAC17, HuCHRAC17, p17, polymerase (DNA directed), epsilon 3 (p17 subunit), YBL1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-147 of human DNA Polymerase epsilon subunit 3 (NP_059139.3). MAERPEDLNLPNAVITRIIKEALPDGVNISKEARSAISRAASVFVLYATSCANNFAMKGKRKTLNASDVLSAMEEMEFQRFVTPLKEALEAYRREQKGKKEASEQKKKDKDKKTDSEEQDKSRDEDNDEDEERLEEEEQNEEEEVDN | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title