missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ DNA Polymerase delta, catalytic subunit Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30106
This item is not returnable.
View return policy
Description
DNA Polymerase delta, catalytic subunit Polyclonal antibody specifically detects DNA Polymerase delta, catalytic subunit in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| DNA Polymerase delta, catalytic subunit | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| CDC2, CDC2 homolog, DNA polymerase delta catalytic subunit, DNA polymerase subunit delta p125, DNA-directed polymerase delta 1, EC 2.7.7.7, POLD, polymerase (DNA directed), delta 1, catalytic subunit (125kD), polymerase (DNA directed), delta 1, catalytic subunit 125kDa | |
| Between 930 and 980 | |
| 100 μg | |
| Breast Cancer, Core ESC Like Genes, DNA Polymerases, DNA Repair, Stem Cell Markers | |
| 5424 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction