missing translation for 'onlineSavingsMsg'
Learn More

DNA Polymerase beta Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£173.00 - £255.00

Specifications

Antigen DNA Polymerase beta
Dilution Western Blot 1:500-1:2000, Immunoprecipitation 1-5uL/mg of lysate, Immunohistochemistry-Paraffin 1:200-1:1000
Applications Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18652256
View Documents
Novus Biologicals
NBP3-05939-100ul
100 μg
£255.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18625205
View Documents
Novus Biologicals
NBP3-05939-50ul
50 μg
£173.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

DNA Polymerase beta Polyclonal antibody specifically detects DNA Polymerase beta in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications

Specifications

DNA Polymerase beta
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
DNA Polymerases, DNA Repair, DNA replication Transcription Translation and Splicing
PBS, pH7.0
5423
IgG
Antigen and protein A Affinity-purified
Western Blot 1:500-1:2000, Immunoprecipitation 1-5uL/mg of lysate, Immunohistochemistry-Paraffin 1:200-1:1000
Polyclonal
Purified
RUO
Human
DNA pol beta, DNA polymerase beta, DNA polymerase beta subunit, EC 2.7.7.7, EC 4.2.99.-, MGC125976, polymerase (DNA directed), beta
Produced in rabbits immunized with E. coli-derived Human DNA Polymerase beta fragment.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.