missing translation for 'onlineSavingsMsg'
Learn More

DNA-PKcs Antibody (3H6), Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18409715 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18409715 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18409715 Supplier Novus Biologicals Supplier No. NBP222128AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DNA-PKcs Monoclonal antibody specifically detects DNA-PKcs in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNA-PKcs
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3H6
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias DNA-dependent protein kinase, DNA-dependent protein kinase catalytic subunit-like, DNA-PK catalytic subunit, DNA-PKcs, DNPK1, EC 2.7.11.1, HYRC, HYRC1, p460
Host Species Mouse
Immunogen Purified recombinant fragment of human DNA PKcs expressed in E. coli. [Uniprot: P78527]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, DNA replication Transcription Translation and Splicing, Non homologous end joining, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 5591
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.