missing translation for 'onlineSavingsMsg'
Learn More

DNA-PKcs Ab-1 Monoclonal Antibody, Epredia™

Inhibit DNA-dependent phosphorylation and perform other studies with Epredia™ DNA-PKcs Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia MS-369-PABX

 View more versions of this product

Product Code. 12681197

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

DNA-dependent Protein Kinase (DNA-PKcs) phosphorylates transcription factors, including Sp1, Oct-1, p53, and SV40 large T antigen in vitro. In vivo, it is involved in repairing double strand DNA breaks.

Host Species: Mouse

Clone: 18-2

Isotype: IgG1

Species Reactivity: Human and Rat. Others not known.

Epitope: aa 1-2713

Immunogen: Human DNA-dependent Protein Kinase purified from HeLa cells

Molecular Weight: 460 kDa

Positive Control: LS174T cells, Tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Inhibits (50%) DNA-dependent Phosphorylation

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

DNA-PKcs Ab-1
Monoclonal
1mg/mL
Mouse
200 μL
DNA Repair
Human, Rat
IgG1
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
18-2
Unconjugated
Human DNA-dependent Protein Kinase purified from HeLa cells
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.