missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNA-PK Recombinant Rabbit Monoclonal Antibody (8O7U9)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558073
This item is not returnable.
View return policy
Description
Immunogen sequence: LKKGGSWIQE INVAEKNWYP RQKICYAKRK LAGANPAVIT CDELLLGHEK APAFRDYVAV ARGSKDHNIR AQEPESGLSE ETQVKCLMDQ ATDPNILGRT.
DNA-PK is a heterotrimeric, DNA-dependent protein kinase that initiates signaling cascades in response to DNA damage. DNA-PK is an atypical kinase, closely related to the PI3 kinases, and is often potently inhibited by small molecules designed to target PI3 kinases. DNA-PK encodes a component of the mediator complex, a transcriptional coactivator complex thought to be required for the expression of almost all genes. The mediator complex is recruited by transcriptional activators or nuclear receptors to induce gene expression, possibly by interacting with RNA polymerase II and promoting the formation of a transcriptional pre-initiation complex. Multiple transcript variants encoding different isoforms have been found for DNA-PK.
Specifications
| DNA-PK | |
| Recombinant Monoclonal | |
| 1.26 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P78527 | |
| PRKDC | |
| A synthetic peptide corresponding to a sequence within amino acids 4021-4120 of human DNA-PKcs (NP_008835.5). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 8O7U9 | |
| Unconjugated | |
| PRKDC | |
| AI326420; AU019811; DNA-dependent protein kinase catalytic subunit; DNAPDcs; DNAPK; DNA-PK catalytic subunit; DNA-PKcs; DNPK1; DOXNPH; doxorubicin nephropathy; dxnph; hyper-radiosensitivity of murine scid mutation, complementing 1; HYRC; HYRC1; I79_017613; IMD26; p350; p460; Prkdc; protein kinase, DNA activated, catalytic polypeptide; protein kinase, DNA-activated, catalytic polypeptide; scid; severe combined immunodeficiency; slip; Xrcc7 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 5591 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction