missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DNA Molecular Weight Marker, Wide Range Ladder (100-12,000 bp)

Product Code. 15480697
Click to view available options
Quantity:
0.5 mL
Unit Size:
0.5mL
This item is not returnable. View return policy

Product Code. 15480697

Brand: Thermo Scientific Chemicals J67051.&I

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

For rapid and precise sizing of DNA strands

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Product Type DNA Molecular Weight Marker/Ladder
Form Liquid
Quantity 0.5 mL
Storage Requirements 2° to 8°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.