missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ DNA Markers
Click to view available options
Quantity:
250 μL
2 x 250 μL
No. of Bands:
9
Concentration:
90 ng/μL
Unit Size:
200 reactions
250µL
Specifications
Specifications
| Concentration | 90 ng/μL |
| Product Type | DNA Molecular Weight Marker/Ladder |
| Form | Liquid |
| For Use With (Application) | Rapid size estimation of PCR products and restriction fragments, Agarose and precast Agarose Gel products |
| Kit Contents | 2 x 250μL DNA marker, 250μL 6X loading Buffer contains Bromophenol Blue |
| No. of Bands | 9 |
| Quantity | 2 x 250 μL |
| Storage Requirements | 4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction