missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DNA Marker, high range, 1,000 Base Pair Ladder

Product Code. p-7082348
Click to view available options
Quantity:
1 mL
2 mL
Unit Size:
1mL
2mL
This item is not returnable. View return policy

Product Code. 15405449

Brand: Thermo Scientific Chemicals J63178.XF

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

DNA ladders are used in gel electrophoresis to determine the size and quantity of testing DNA fragments of genomic, plasmid, and PCR DNA. It is commonly used in the studies of plasmid DNA and PCR products. 1 kb DNA Ladder has been used as a molecular marker to determine the molecular weight and size of double stranded DNA during electrophoresis. 1 kb Ladder contains 11 fragments consisting of 500 bp repeats from 0.5 to 3 kb, 1 kb repeats from 3 to 6 kb, and 2 kb repeats from 6 to 10 kb. In NA electrophoresis, five ?l of the marker should be diluted in gel loading buffer and then loaded in a single lane on an agarose or polyacrylamide gel. Suitable for use in Northern and Southern blotting. All DNA markers can be stained with ethidium bromide, SYBR™ Green, and Nancy-520.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Molecular Weight Marker
Form Liquid
Quantity 1 mL
Storage Requirements −30° to −10°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.