missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNA Ligase IV Recombinant Rabbit Monoclonal Antibody (23GB2515), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | DNA Ligase IV |
|---|---|
| Clone | 23GB2515 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089769
![]() |
Invitrogen™
MA553494 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088499
|
Invitrogen™
MA553495 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DNA Ligase IV Recombinant Monoclonal antibody specifically detects DNA Ligase IV in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| DNA Ligase IV | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P49917 | |
| 3981 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB2515 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 5830471N16Rik; DNA joinase; DNA ligase (ATP) 4; DNA ligase 4; DNA ligase IV; DNA repair enzyme; Lig4; LIG4S; ligase IV, DNA, ATP-dependent; polydeoxyribonucleotide synthase [ATP] 4; polynucleotide ligase; sealase; tiny | |
| A synthesized peptide derived from human DNA Ligase IV (504-685AA). | |
| Primary | |
| -20°C | |
| LIG4 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title