missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA Ligase IV Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35747-20ul
This item is not returnable.
View return policy
Description
DNA Ligase IV Polyclonal antibody specifically detects DNA Ligase IV in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| DNA Ligase IV | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| DNA joinase, DNA ligase 4, DNA ligase IV, DNA repair enzyme, EC 6.5.1.1, ligase IV, DNA, ATP-dependent, Polydeoxyribonucleotide synthase [ATP] 4, polynucleotide ligase, sealase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-210 of human DNA Ligase IV (NP_002303.2).,, Sequence:, MAASQTSQTVASHVPFADLCSTLERIQKSKGRAEKIRHFREFLDSWRKFHDALHKNHKDVTDSFYPAMRLILPQLERERMAYGIKETMLAKLYIELLNLPRDGKDALKLLNYRTPTGTHGDAGDFAMIAYFVLKPRCLQKGSLTIQQVNDLLDSIASNNSAKRKDLIKKSLLQLITQSSALEQKWLIRMIIKDLKLGVSQQTIFSVFHND | |
| 20 μL | |
| Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Non homologous end joining | |
| 3981 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction