missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNA Ligase III Recombinant Rabbit Monoclonal Antibody (HL2280)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550473
This item is not returnable.
View return policy
Description
DNA Ligase III Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
There are four distinct ligase proteins found in mammalian cells. The activity of these DNA ligases is required for the joining of DNA strand breaks that occur during DNA replication, repair and recombination. DNA Ligase III gene product migrates as a 100 kDa (a-form) protein in somatic tissues, is ubiquitously expressed and complexes with XRCC-1 and a 97 kDa (beta form) protein that is found in testis. DNA Ligase III is involved in rejoining DNA single-strand breaks and in the base excision repair pathway.
Specifications
| DNA Ligase III | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P49916, P97386 | |
| LIG3 | |
| Recombinant fragment of human DNA ligase III. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| HL2280 | |
| Unconjugated | |
| LIG3 | |
| D11Wsu78e; DNA ligase (ATP) 3; DNA ligase 3; DNA ligase III; hypothetical protein; LIG2; LIG3; ligase II, DNA, ATP-dependent; ligase III, DNA, ATP-dependent; polydeoxyribonucleotide synthase [ATP] 3; Unknown (protein for MGC:134139) | |
| Rabbit | |
| Protein A | |
| RUO | |
| 16882, 303369, 3980 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction