missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA Ligase I Mouse anti-Human, Mouse, Rat, Bovine, Clone: 1A9, Novus Biologicals™
Mouse Monoclonal Antibody
£377.00
Specifications
| Antigen | DNA Ligase I |
|---|---|
| Clone | 1A9 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1 to 2 μg/mL, ELISA, Immunoprecipitation |
| Applications | Western Blot, ELISA, Immunoprecipitation |
Description
DNA Ligase I Monoclonal specifically detects DNA Ligase I in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, ELISA, Immunoprecipitation.Specifications
| DNA Ligase I | |
| 0.2 mg/mL | |
| Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Base Excision Repair, Cancer, DNA Repair, Nucleotide Excision Repair, Tumor Suppressors | |
| 10 mM PBS with 0.05% BSA | |
| 3978 | |
| IgG1 κ | |
| Protein A or G purified |
| 1A9 | |
| Western Blot 1 to 2 μg/mL, ELISA, Immunoprecipitation | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Bovine | |
| DNA ligase 1, DNA ligase I, EC 6.5.1.1, ligase I, DNA, ATP-dependent, MGC117397, MGC130025, Polydeoxyribonucleotide synthase [ATP] 1, polydeoxyribonucleotide synthase ATP 1 | |
| Full-length native cow LDNA Ligase I protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title