missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA Ligase I Antibody (7B2), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26168-50ul
This item is not returnable.
View return policy
Description
DNA Ligase I Monoclonal antibody specifically detects DNA Ligase I in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| DNA Ligase I | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| 7B2 | |
| Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 | |
| DNA ligase 1, DNA ligase I, EC 6.5.1.1, ligase I, DNA, ATP-dependent, MGC117397, MGC130025, Polydeoxyribonucleotide synthase [ATP] 1, polydeoxyribonucleotide synthase ATP 1 | |
| A synthesized peptide derived from Human DNA Ligase I [UniProt P18858] | |
| 50 μL | |
| Apoptosis, Base Excision Repair, Cancer, DNA Repair, Nucleotide Excision Repair, Tumor Suppressors | |
| 3978 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction