missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA Ligase I Antibody (1A9), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08267AF350
This item is not returnable.
View return policy
Description
DNA Ligase I Monoclonal antibody specifically detects DNA Ligase I in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
| DNA Ligase I | |
| Monoclonal | |
| Alexa Fluor 350 | |
| DNA ligase 1, DNA ligase I, EC 6.5.1.1, ligase I, DNA, ATP-dependent, MGC117397, MGC130025, Polydeoxyribonucleotide synthase [ATP] 1, polydeoxyribonucleotide synthase ATP 1 | |
| Full-length native bovine DNA Ligase I protein (Uniprot: P18858) | |
| 100 μg | |
| Apoptosis, Base Excision Repair, Cancer, DNA Repair, Nucleotide Excision Repair, Tumor Suppressors | |
| 3978 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, Immunoprecipitation | |
| 1A9 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Bovine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction