missing translation for 'onlineSavingsMsg'
Learn More

DNA Ladder, 100 kb Plus, Thermo Scientific™

Product Code. 15815238 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 μL
Unit Size:
500µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15815238 500 μL 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15815238 Supplier Thermo Scientific Supplier No. J76714.Y^

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

6X Loading Buffer consists of bromophenol blue, xylene cyanol FF and Ficoll™

 

  • Set of 14 easy-to remember band sizes for use in qualitative agarose gel analysis ranging in size from 12 kb down to 100 bp
  • Two high intensity reference bands of 4 kb and 500 bp are included
  • Recommended amount (5μL) per lane
  • Supplied with 6X Loading Buffer consists of bromophenol blue, xylene cyanol FF and Ficoll for convenience to prepare DNA samples for agarose gel electrophoresis
  • Tracking dyes do not interfere with UV illumination of ethidium bromide stained gels
  • Fragments: 12 kb; 10 kb; 8 kb; 6 kb; 4 kb; 3 kb; 2 kb; 1.5 kb; 1 kb; 500 bp; 400 bp; 300 bp; 200 bp; 100 bp
TRUSTED_SUSTAINABILITY

Specifications

Buffer 6 x DNA Loading Buffer (OXG)
Color Blue, Orange
Concentration 6 x Loading Dye
Form Liquid
Quantity 500 μL
Storage Requirements 4°C
Product Type DNA Molecular Weight Marker
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.