missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA helicase HEL308 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92705-0.1ml
This item is not returnable.
View return policy
Description
DNA helicase HEL308 Polyclonal antibody specifically detects DNA helicase HEL308 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| DNA helicase HEL308 | |
| Polyclonal | |
| Western Blot 1:1000-1:3000 | |
| EC 3.6.1, EC 3.6.4.12, Hel308, HEL308DNA helicase HEL308, helicase POLQ-like, helicase, POLQ-like, MGC20604, Mus308-like helicase, POLQ-like helicase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human DNA helicase HEL308 (NP_598375.3). MDECGSRIRRRVSLPKRNRPSLGCIFGAPTAAELVPGDEGKEEEEMVAENRRRKTAGVLPVEVQPLLLSDSPECLVLGGGDTNPDLLRHMPTDRGVGDQPNDSEVDMFGDYDSFTENSFIAQVDDLEQKYMQLPEHKKHATDFATENLCSESIKNKLSITTIGNLTELQTDKHTENQSGYEGVTIEPGADLLYDVPSSQAIYFENLQNSSNDLGDHSMKERDWKSSSHNTVNEELPHNCIEQPQQNDESSSKVRTSSDMNRRKSIKDHLK | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 113510 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction