missing translation for 'onlineSavingsMsg'
Learn More

Extract All DNA Extract All Reagents Kit

Product Code. 11551296 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
5 mL
20 mL
200 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11551296 200 mL Each
11571296 5 mL Each
11561296 20 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11551296 Supplier Extract All Supplier No. 4402599

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Provides PCR-ready DNA from a wide variety of sample types, ranging from blood to buccal swabs to plant tissues, in 5 minutes

Simply mix your sample with the DNA lysis solution provided. Incubate for 3 minutes and add the DNA stabilizing solution, also provided.

After extraction, robust PCR amplification can be completed using TaqMan™ GTXpress™ Master Mix or TaqMan™ OpenArray™ Genotyping Master Mix (pre-amplified sample only), depending on your application. PCR can be performed using either Fast or Standard mode thermal cycling conditions. The DNA Extract All Reagents Kit is included in the TaqMan Sample-to-SNP Kit, so workflow details can be found in the TaqMan™ Sample-to-SNP user guide.

Thermal cycling conditions should be followed according to the master mix being used. TaqMan™ GTXpress™ Master Mix is required for PCR amplification of sample prepared using the DNA Extract All Reagents Kit. If sample has undergone preamplification, OpenArray™ Genotyping Master Mix may also be used.

Recommended Storage

Store at 2° to 8°C.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 2–8°C.
Product Line Extract All
Quantity 200 mL
Reagent Type Cell Lysis Buffer
Test Time 5 min
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.