missing translation for 'onlineSavingsMsg'
Learn More

DNA Antibody (11B6), Novus Biologicals™

Product Code. 18065434 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18065434 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18065434 Supplier Novus Biologicals Supplier No. NB1001647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DNA Monoclonal specifically detects DNA in All Species samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNA
Applications ELISA
Classification Monoclonal
Clone 11B6
Concentration LYOPH
Conjugate Unconjugated
Dilution ELISA
Gene Alias Double stranded deoxyribonucleic acid, Double stranded DNA, dsDNA
Host Species Mouse
Immunogen calf dsDNA
Purification Method Unpurified
Quantity 1 mL
Regulatory Status RUO
Research Discipline Chromatin Research
Primary or Secondary Primary
Test Specificity In ELISA : This shows 100% reactivity to calf dsDNA and 5% to calf ssDNA. It does not react with RNA.
Reconstitution Reconstitute with 1.0 ml distilled water.
Target Species All species
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Supernatant
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.