missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DMXL2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24415-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a protein with 12 WD domains. Proteins with WD domains are involved in many functions including participation in signal transduction pathways. Participation of the encoded protein in regulation of the Notch signaling pathway has been demonstrated in vitro using several human cell lines. A gene encoding a similar protein is located on chromosome 5. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
| DMXL2 | |
| Polyclonal | |
| Unconjugated | |
| DMXL2 | |
| Dmx like 2, DmX like protein 2, DMXL2, KIAA0856, Rabconnectin 3, RC3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23312, 235380 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8BPN8, Q8TDJ6 | |
| DMXL2 | |
| DMXL2 Fusion Protein Ag19720 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction