missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DMX-5804, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
DMX-5804 is a potent, orally active and selective MAP4K4 inhibitor, with an IC50 of 3 nM, a pIC50 of 8.55 for human MAP4K4, less potent on MINK1/MAP4K6 (pIC50, 8.18), and TNIK/MAP4K7 (pIC50, 7.96). DMX-5804 enhances cardiomyocyte survival, and reduces ischemia-reperfusion injury in mice.
Specifications
Specifications
| Chemical Name or Material | DMX-5804 |
| CAS | 2306178-56-1 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C21H19N3O3 |
| Quantity | 50 mg |
| Solubility Information | DMSO : 125 mg/mL (345.89 mM; Need ultrasonic) |
| SMILES | O=C1NC=NC2=C1C(C3=CC=C(OCCOC)C=C3)=CN2C4=CC=CC=C4 |
| Molecular Weight (g/mol) | 361.39 |
| Formula Weight | 361.39 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction