missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DMWD Monoclonal Antibody (3F5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA518749
This item is not returnable.
View return policy
Description
Peptide Sequence: GGEKPSGPVP RSRLDPAKVL GTALCPRIHE VPLLEPLVCK KIAQERLTVL LFLEDCIITA CQEGLICTWA RPGKAGISSQ PGNSPSGTVV.
DMWD (DM1 Locus, WD Repeat Containing) is a Protein Coding gene. Diseases associated with DMWD include Myotonic Dystrophy 1 and Corticosteroid Allergy. An important paralog of this gene is WDR20. [GeneCards].
Specifications
| DMWD | |
| Monoclonal | |
| 0.2-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q09019 | |
| Dmwd | |
| DMWD (AAH19266, 245 a.a. ~ 334 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot | |
| 3F5 | |
| Unconjugated | |
| Dmwd | |
| D19S593E; Dm9; DMRN9; DMR-N9; DMWD; dystrophia myotonica linked gene, N9; dystrophia myotonica WD repeat-containing protein; dystrophia myotonica, WD repeat containing; dystrophia myotonica-containing WD repeat motif; dystrophia myotonica-containing WD repeat motif protein; gene59; Protein 59; Protein DMR-N9 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1762 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction