missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DMRTC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599788
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total DMRTC2.
DMRTC2 belongs to the DMRT family.It may be involved in sexual development.
Specifications
| DMRTC2 | |
| Polyclonal | |
| Unconjugated | |
| DMRTC2 | |
| 4933432E21Rik; DMRT like family C2; Dmrt7; DMRTC2; DMRT-like family C2; doublesex and mab-3 related transcription factor 7; doublesex and mab-3 related transcription factor like family C2; Doublesex- and mab-3-related transcription factor 7; Doublesex- and mab-3-related transcription factor C2; Unknown (protein for MGC:133756) | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 63946 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8IXT2 | |
| DMRTC2 | |
| A synthesized peptide derived from human DMRTC2(Accession Q8IXT2), corresponding to amino acid residues M1-T41. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto